Order Entry
Singapore
ContactUsLinkComponent
NANOCOLOR Accessory set for TOC (big) content: 1 magnetic stirrer (15 stirr positions) 6 beakers, 100 mL 6 magnetic stir bars 1 * 1 SET
Catalog # MANA916991
Supplier:  MACHEREY-NAGEL
undefined
NANOCOLOR Accessory set for TOC (big) content: 1 magnetic stirrer (15 stirr positions) 6 beakers, 100 mL 6 magnetic stir bars 1 * 1 SET
Catalog # MANA916991
Supplier:  MACHEREY-NAGEL