Test papers, ion
Cyanogen Check (CN-), 7.0 – 9.0, 0 – 200 mg/l
Catalog # ADVT07030100
Supplier: Advantech
Jump to:
Product Details & Documents
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
Packaging: Strips, 5×85 mm.
Specifications
- pH range:7.0 – 9.0
- Description:Cyanogen Check (CN-)
- Seq:06
- Range:0 – 200 mg/l