Test papers, ion
Nickel Check (Ni2+), 2.0 – 7.0, 0 − 50 mg/l
Catalog # ADVT07030070
Supplier: Advantech
Jump to:
Product Details & Documents
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
Packaging: Strips, 5×85 mm.
Specifications
- pH range:2.0 – 7.0
- Description:Nickel Check (Ni2+)
- Seq:05
- Range:0 − 50 mg/l