Test papers, ion
Iron Check (Fe2+), 1.0 – 6.0, 0 – 500 mg/l
ADVT07030060
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
Packaging: Strips, 5×85 mm.
- pH range:1.0 – 6.0
- Description:Iron Check (Fe2+)
- Seq:04
- Range:0 – 500 mg/l