Test papers, ion
Chrome Check-A (Cr6+), 1.0 – 9.0, 0 – 100 mg/l
Catalog # ADVT07030051
Supplier: Advantech
Jump to:
Product Details & Documents
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
Packaging: Strips, 5×85 mm.
Specifications
- pH range:1.0 – 9.0
- Description:Chrome Check-A (Cr6+)
- Seq:08
- Range:0 – 100 mg/l