Test papers, ion
Chrome Check (Cr6+), 1.0 – 4.0, 0 – 1000 mg/l
ADVT07030041
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
: Strips, 5×85 mm.
- pH range:1.0 – 4.0
- Description:Chrome Check (Cr6+)
- Seq:07
- Range:0 – 1000 mg/l