Test papers, ion
Copper Check (Cu), 3.0 – 5.0, 0 – 1000 mg/l
Catalog # ADVT07030020
Supplier: Advantech
Jump to:
Product Details & Documents
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
Packaging: Strips, 5×85 mm.
Specifications
- pH range:3.0 – 5.0
- Description:Copper Check (Cu)
- Seq:02
- Range:0 – 1000 mg/l