Test papers, ion
Aluminium Check (Al3+), 3.0 – 6.0, 0 − 50 mg/l
Catalog # ADVT07030010
Supplier: Advantech
Jump to:
Product Details & Documents
Detects concentration of specified ion in solution. Two-point indicator strip confirms within pH range and ion concentration by color comparison.
Dip the indicator test strip in solution and compare to the color chart on the container.
Packaging: Strips, 5×85 mm.
Specifications
- pH range:3.0 – 6.0
- Description:Aluminium Check (Al3+)
- Seq:01
- Range:0 − 50 mg/l