pH indicator papers
pH indicator paper, BTB, roll, 6,2 – 7,8
ADVT07011020
Various types of indicator papers cover a wide pH range to allow for good accuracy of determining pH value of a solution.
High sensitivity and sharp color change. Narrow pH unit graduation: 0,2 – 0,4 (UNIV/ WR: 1,0 – 2,0 units).
: Booklet of 20 strips, 7×70 mm; Roll, 9 mm wide in dispenser.
- pH range:6,2 – 7,8
- Description:pH indicator paper, BTB, roll
- Seq:16