pH indicator papers
pH indicator paper, number 20, booklet, 5,0 – 8,0
Catalog # ADVT07010110
Supplier: Advantech
Jump to:
Product Details & Documents
Various types of indicator papers cover a wide pH range to allow for good accuracy of determining pH value of a solution.
High sensitivity and sharp color change. Narrow pH unit graduation: 0,2 – 0,4 (UNIV/ WR: 1,0 – 2,0 units).
: Booklet of 20 strips, 7×70 mm; Roll, 9 mm wide in dispenser.
Specifications
- pH range:5,0 – 8,0
- Description:pH indicator paper, number 20, booklet
- Seq:11