Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DIS-300-S-M.50 Tube with dispersing element, 300 ml, 50 Pcs., stainless steel 1 * 50 items
Catalog # IKAA20017313
Supplier:
IKA
Expand image
DIS-300-S-M.50 Tube with dispersing element, 300 ml, 50 Pcs., stainless steel 1 * 50 items
Catalog # IKAA20017313
Supplier:
IKA