Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BDH BUFFER REF STD PH7 YELLOW 4L 1 * 4 L
Catalog # BDHB5048-4L
Supplier:
BDH US PRODUCED
Certificates
Expand image
BDH BUFFER REF STD PH7 YELLOW 4L 1 * 4 L
Catalog # BDHB5048-4L
Supplier:
BDH US PRODUCED
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation