Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Porapak Q 100/120 mesh 6’ x 1/8” od SS for Shimadzu GC-2014ATF with FID+TCD Detector 1 * 1 items
HISMC5000
:
HICHROM
Porapak Q 100/120 mesh 6’ x 1/8” od SS for Shimadzu GC-2014ATF with FID+TCD Detector 1 * 1 items
HISMC5000
:
HICHROM