Order Entry
Singapore
ContactUsLinkComponent
Violet Red Bile Glucose Agar, granulated is recommended for detection and enumeration of Enterobacteriaceae from pharmaceutical products in accordance with the microbial limit testing by harmonized methodology of USP/EP/BP/JP/IP. 1 * 100 g
Catalog # VWRC85949.0100SAMP
Supplier:  VWR Chemicals
undefined
Violet Red Bile Glucose Agar, granulated is recommended for detection and enumeration of Enterobacteriaceae from pharmaceutical products in accordance with the microbial limit testing by harmonized methodology of USP/EP/BP/JP/IP. 1 * 100 g
Catalog # VWRC85949.0100SAMP
Supplier:  VWR Chemicals
Related Information