Order Entry
Singapore
ContactUsLinkComponent
Plate Count Agar is recommended for the determination of plate counts of microorganisms in food, water,waste water and clinical samples. 1 * 100 g
Catalog # VWRC85941.0100SAMP
Supplier:  VWR Chemicals
undefined
Plate Count Agar is recommended for the determination of plate counts of microorganisms in food, water,waste water and clinical samples. 1 * 100 g
Catalog # VWRC85941.0100SAMP
Supplier:  VWR Chemicals
Related Information