Order Entry
Singapore
ContactUsLinkComponent
VWR® Critical Swab® Elongated Foam Tipped Swab, Polypropylene Handle, 108.2mm long, 3.6mm in diameter 1 * 500 items
Catalog # VWRU89031-284
Supplier:  VWR Collection
VWR® Critical Swab® Elongated Foam Tipped Swab, Polypropylene Handle, 108.2mm long, 3.6mm in diameter 1 * 500 items
Catalog # VWRU89031-284
Supplier:  VWR Collection