Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
CONDUCTIVITY STANDARD, 84 µS/CM AT 25°C 1 * 500 mL
RICCR2236210-500A
:
Ricca Chemical
CONDUCTIVITY STANDARD, 84 µS/CM AT 25°C 1 * 500 mL
RICCR2236210-500A
:
Ricca Chemical