Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
OSMOLALITY STANDARD/700MOSM/KG H2O 1 * 1 KIT
REGERE-OSM-700
:
REAGECON
OSMOLALITY STANDARD/700MOSM/KG H2O 1 * 1 KIT
REGERE-OSM-700
:
REAGECON