Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
EC HPLC column EC 150/3 NUCLEOSIL 100-5 C18 length: 150 mm, ID: 3 mm pack of 1 1 * 1 items
MANA720120.30
:
MACHEREY-NAGEL
EC HPLC column EC 150/3 NUCLEOSIL 100-5 C18 length: 150 mm, ID: 3 mm pack of 1 1 * 1 items
MANA720120.30
:
MACHEREY-NAGEL