Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Buffer Solution, pH 7.00, Color-Coded Yellow, 500ml 1 * 1 items
Catalog # XXXXSB107-500
Supplier:
Chemicals
Expand image
Buffer Solution, pH 7.00, Color-Coded Yellow, 500ml 1 * 1 items
Catalog # XXXXSB107-500
Supplier:
Chemicals