Order Entry
Singapore
ContactUsLinkComponent
Ubbelohde Viscometer, Calibrated, Size:50, Range:0.8 to 4 cSt, Approx constant:0.004 cSt/s 1 * 1 items
Catalog # CANN9721-K53
Supplier:  CANNON INSTRUMENT
Ubbelohde Viscometer, Calibrated, Size:50, Range:0.8 to 4 cSt, Approx constant:0.004 cSt/s 1 * 1 items
Catalog # CANN9721-K53
Supplier:  CANNON INSTRUMENT