D-(+)-Galactose ≥99%
Plastic bottle, 5 kg
Catalog # ALFAJ15995.A7
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: D-Galactose, (2R,3S,4S,5R)-2,3,4,5,6-Pentahydroxyhexanal, (2R,3S,4S,5R)-Pentahydroxyhexanal
- CAS Number:59-23-4
- Formula:C₆H₁₂O₆
- MDL Number:MFCD00151230
- Molecular Weight:180.16 g/mol
- Merck Index:14,04335
- Melting Pt:164…168 °C
- Density:1,50
- Storage Temperature:Ambient
- EINECS:200-416-4
Specifications
Specification Test Results
- Assay (HPLC):≥99.0%
- Identification (FTIR):Conforms
- Residue on Ignition:≤0.5%
- Loss on Drying:≤0.5%
- Optical Rotation (c=4 in H₂O and NH₃):≥77°
Specifications
- Pack type:Plastic bottle
- Pk:5 kg



