Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
pH BUFFERS pH7 YELLOW 1 * 500 mL
Catalog # RICCR1551000-500A
Supplier:
Ricca Chemical
Certificates
Expand image
pH BUFFERS pH7 YELLOW 1 * 500 mL
Catalog # RICCR1551000-500A
Supplier:
Ricca Chemical
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation