Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
HAMS F12 MEDIUM WITH STABLE GLUTAMINE WITH 1.176 G/L NAHCO3 1 * 500 mL
Catalog # PANBP04-15500
Supplier:
PAN BIOTECH
Expand image
HAMS F12 MEDIUM WITH STABLE GLUTAMINE WITH 1.176 G/L NAHCO3 1 * 500 mL
Catalog # PANBP04-15500
Supplier:
PAN BIOTECH