Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Kjeldahl Catalyst (Cu) (10.26% in CuSO4 5H2O) tablets 1 * 1.000 items
Catalog # APPC177040.1246
Supplier:
PanReac AppliChem
Expand image
Kjeldahl Catalyst (Cu) (10.26% in CuSO4 5H2O) tablets 1 * 1.000 items
Catalog # APPC177040.1246
Supplier:
PanReac AppliChem