Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Pan Size: 17.25 x 13.50 x 4.96 (43.8cm x 34.3cm x 12.63cm) Volume: 9 Liter 1 * 1 items
DIVEIPAN-3000
:
DIVERSIFIED BIOTECH
Pan Size: 17.25 x 13.50 x 4.96 (43.8cm x 34.3cm x 12.63cm) Volume: 9 Liter 1 * 1 items
DIVEIPAN-3000
:
DIVERSIFIED BIOTECH