Bulk sorbents
Bulk silica C₈, 100 g
HONC9942
Wide selection of support materials.
- Every stationary phase batch tested to ensure lot-to-lot reproducibility
- Reliable quality
- Pk:100 g
- Description:Bulk silica C₈
Wide selection of support materials.