Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
pH electrolyte (KCl saturated AgCl, 30ml) 1 * 30 mL
Catalog # OHAU30059255
Supplier:
OHAUS
Expand image
pH electrolyte (KCl saturated AgCl, 30ml) 1 * 30 mL
Catalog # OHAU30059255
Supplier:
OHAUS