Skip to Header
Skip to Main Content
Skip to Footer
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Buffer Solution pH 1.68 , 250 mL 1 * 250 mL
Buffer Solution pH 1.68 , 250 mL 1 * 250 mL
Print
Buffer Solution pH 1.68 , 250 mL 1 * 250 mL
Catalog # OHAU30100424
Supplier:
OHAUS
Buffer Solution pH 1.68 , 250 mL 1 * 250 mL
Catalog # OHAU30100424
Supplier:
OHAUS
Expand image
Return to Top
Buffer Solution pH 1.68 , 250 mL 1 * 250 mL
Add to cart
Price
Inquire For Price
Log in
for availability
Unit of Measure
1 * 250 mL
-
+
Inquire For Price
Return Policy
Product Details & Documents