$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
COLOR STANDARD, APHA/HAZEN (PLATINUM- CO 1 * 500 mL
RICCR2230050-500C
:
Ricca Chemical
COLOR STANDARD, APHA/HAZEN (PLATINUM- CO 1 * 500 mL
RICCR2230050-500C
:
Ricca Chemical