$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Singapore
ContactUsLinkComponent
Color Standard, APHA/Hazen (Platinum-Cobalt), Color= 5 1 * 500 mL
RICCR2230005-500C
:
Ricca Chemical
Color Standard, APHA/Hazen (Platinum-Cobalt), Color= 5 1 * 500 mL
RICCR2230005-500C
:
Ricca Chemical