Order Entry
Singapore
ContactUsLinkComponent
Automation Tips, 10µL, Sterile, Liquid Level Sensing,Non-Filtered, Nested Tip Rack Stack. For STAR, STARlet, STARplus and Nimbus®. 96 tips per rack. 1 * 4 items
Catalog # AXYGHT-10-CBK-STKS
Supplier:  Corning
Automation Tips, 10µL, Sterile, Liquid Level Sensing,Non-Filtered, Nested Tip Rack Stack. For STAR, STARlet, STARplus and Nimbus®. 96 tips per rack. 1 * 4 items
Catalog # AXYGHT-10-CBK-STKS
Supplier:  Corning