Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TISSUE HOMOGENIZER POTTER-ELEHJEM 1 * 2 items
Catalog # ACEG8355-10
Supplier:
Ace Glass
Expand image
TISSUE HOMOGENIZER POTTER-ELEHJEM 1 * 2 items
Catalog # ACEG8355-10
Supplier:
Ace Glass