Cylinders, borosilicate glass, with hexagonal base
25 ml
Catalog # SCOT213911472
Supplier: DWK Life Sciences
Cylinders, borosilicate glass, with hexagonal base
Catalog # SCOT213911472
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
DURAN® borosilicate glass 3.3, clear.
- Without graduation
- Hexagonal base
- Base with three knobs which increase its stability
: DIN EN ISO 4788.
Specifications
- Capacity:25 ml
- Seq:03