Order Entry
Singapore
ContactUsLinkComponent
Package includes laboratory stirrer EUROSTAR 100 control, R 1345 Propeller stirrer, 4-bladed, R 2723 Telescopic stand, R 270 Boss head clamp, RH 5 Strap clamp, labworldsoft 1 * 1 items
Catalog # IKAA10000359
Supplier:  IKA
Package includes laboratory stirrer EUROSTAR 100 control, R 1345 Propeller stirrer, 4-bladed, R 2723 Telescopic stand, R 270 Boss head clamp, RH 5 Strap clamp, labworldsoft 1 * 1 items
Catalog # IKAA10000359
Supplier:  IKA