ChemiDoc-It 615 Cam/Dkrm Kit615 camera: 3.2 mp, 16 bit monochrome, 16 File Bit Depth (A/D), digital camera and mount, with binning c apability. Peltier cooling to -60C from ambient, MOTORISED fixed 25mm, f/0.95 lens, close up diopter, USB 2.0 connec 1 * 1 items
Catalog # UVPA97-0726-02
Supplier: UVP ULTRA VIOLET PRODUCTS