ChemiDoc-It 2 815 Cam/Dkrm Kit815 camera: 8.1 mp, 16 bit monochrome, digital camera with mount, with binning capabilities, cooling t o -57C from ambient. MOTORISED fixed 0.25mm, f/0.95 lens, close up diopter, 50x50mm Ethidium Bromide filter, USB car 1 * 1 items
Catalog # UVPA97-0722-02
Supplier: UVP ULTRA VIOLET PRODUCTS