Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Singapore
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SODIUM AZIDE 0.5% (W/V) AQUEOUS SOLUTION 1 * 500 mL
Catalog # RICCR7144000-500A
Supplier:
Ricca Chemical
Expand image
SODIUM AZIDE 0.5% (W/V) AQUEOUS SOLUTION 1 * 500 mL
Catalog # RICCR7144000-500A
Supplier:
Ricca Chemical