Order Entry
Singapore
ContactUsLinkComponent
PEROXIDASE-CONJUGATED GOAT Fab FRAGMENT TO RABBIT IgG (WHOLE MOLECULE). Anti-RabbitHost: goatForm: lyophilized (Fab)Applications: Im munostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.HRP Conjugated 1 * 2 mL
Catalog # ICNA0855688
Supplier:  MP Biomedicals
PEROXIDASE-CONJUGATED GOAT Fab FRAGMENT TO RABBIT IgG (WHOLE MOLECULE). Anti-RabbitHost: goatForm: lyophilized (Fab)Applications: Im munostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.HRP Conjugated 1 * 2 mL
Catalog # ICNA0855688
Supplier:  MP Biomedicals