Order Entry
Singapore
ContactUsLinkComponent
PEROXIDASE - CONJUGATED SHEEP IgG FRACTION TO RABBIT IgG (WHOLE MOLECULE). Anti-Rabbit Host: sheep Form: lyophilized (IgG) Applicati ons: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855677
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED SHEEP IgG FRACTION TO RABBIT IgG (WHOLE MOLECULE). Anti-Rabbit Host: sheep Form: lyophilized (IgG) Applicati ons: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855677
Supplier:  MP Biomedicals