Order Entry
Singapore
ContactUsLinkComponent
RHODAMINE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG (WHOLE MOLECULE). Anti-Rabbit Host: goat Form: affinity purifie d liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855674
Supplier:  MP Biomedicals
RHODAMINE - CONJUGATED GOAT AFFINITY PURIFIED ANTIBODY TO RABBIT IgG (WHOLE MOLECULE). Anti-Rabbit Host: goat Form: affinity purifie d liquid Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855674
Supplier:  MP Biomedicals