Order Entry
Singapore
ContactUsLinkComponent
GOAT IgG FRACTION TO RABBIT IgM (µ CHAIN). Anti-RabbitHost: goatForm: lyophilized (IgG)Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855627
Supplier:  MP Biomedicals
GOAT IgG FRACTION TO RABBIT IgM (µ CHAIN). Anti-RabbitHost: goatForm: lyophilized (IgG)Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 5 mL
Catalog # ICNA0855627
Supplier:  MP Biomedicals