Order Entry
Singapore
ContactUsLinkComponent
BIOTIN-CONJUGATED SHEEP AFFINITY PURIFIED F(ab')2 FRAGMENT TO MOUSE IgG (NO CROSS TO HUMAN). Anti-MouseHost: sheepForm: affinity pur ified F(ab)2 liquidApplications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue 1 * 1 mL
Catalog # ICNA0855600
Supplier:  MP Biomedicals
BIOTIN-CONJUGATED SHEEP AFFINITY PURIFIED F(ab')2 FRAGMENT TO MOUSE IgG (NO CROSS TO HUMAN). Anti-MouseHost: sheepForm: affinity pur ified F(ab)2 liquidApplications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue 1 * 1 mL
Catalog # ICNA0855600
Supplier:  MP Biomedicals