Order Entry
Singapore
ContactUsLinkComponent
FLUORESCEIN-CONJUGATED GOAT IgG FRACTION TO MOUSE IgG Fc. Anti-MouseHost: goatForm: lyophilized (IgG).Applications: Immunostaining o f acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mL
Catalog # ICNA0855497
Supplier:  MP Biomedicals
FLUORESCEIN-CONJUGATED GOAT IgG FRACTION TO MOUSE IgG Fc. Anti-MouseHost: goatForm: lyophilized (IgG).Applications: Immunostaining o f acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections.FITC Conjugated 1 * 2 mL
Catalog # ICNA0855497
Supplier:  MP Biomedicals