GOAT AFFINITY PURIFIED F(ab')2 FRAGMENT TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: affinity purified liquid F(ab ')2 Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855487
GOAT AFFINITY PURIFIED F(ab')2 FRAGMENT TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: goat Form: affinity purified liquid F(ab ')2 Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855487