Order Entry
Singapore
ContactUsLinkComponent
GOAT AFFINITY PURIFIED ANTIBODY TO MOUSE IgM (µ CHAIN). Anti-Mouse Host: goat Form: affinity purified liquid Applications: Immunosta ining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855484
Supplier:  MP Biomedicals
GOAT AFFINITY PURIFIED ANTIBODY TO MOUSE IgM (µ CHAIN). Anti-Mouse Host: goat Form: affinity purified liquid Applications: Immunosta ining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mg
Catalog # ICNA0855484
Supplier:  MP Biomedicals