Order Entry
Singapore
ContactUsLinkComponent
RABBIT IgG FRACTION TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: rabbit Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855456
Supplier:  MP Biomedicals
RABBIT IgG FRACTION TO MOUSE IgG (WHOLE MOLECULE). Anti-Mouse Host: rabbit Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855456
Supplier:  MP Biomedicals