Order Entry
Singapore
ContactUsLinkComponent
FLUORESCEIN - CONJUGATED RABBIT IgG FRACTION TO HORSE IgG (WHOLE MOLECULE). Anti-Horse Host: rabbit Form: lyophilized (IgG) Applicat ions: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855415
Supplier:  MP Biomedicals
FLUORESCEIN - CONJUGATED RABBIT IgG FRACTION TO HORSE IgG (WHOLE MOLECULE). Anti-Horse Host: rabbit Form: lyophilized (IgG) Applicat ions: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. FITC Conjugated 1 * 2 mL
Catalog # ICNA0855415
Supplier:  MP Biomedicals