Order Entry
Singapore
ContactUsLinkComponent
RABBIT ANTISERUM TO DOG IgG (WHOLE MOLECULE). Anti-Dog Host: rabbit Form: lyophilized Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855311
Supplier:  MP Biomedicals
RABBIT ANTISERUM TO DOG IgG (WHOLE MOLECULE). Anti-Dog Host: rabbit Form: lyophilized Applications: Immunostaining of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. 1 * 2 mL
Catalog # ICNA0855311
Supplier:  MP Biomedicals