Order Entry
Singapore
ContactUsLinkComponent
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO HUMAN IgG Fab. Anti-Human Host: goat Form: lyophilized (IgG) Applications: Immunostaini ng of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855223
Supplier:  MP Biomedicals
PEROXIDASE - CONJUGATED GOAT IgG FRACTION TO HUMAN IgG Fab. Anti-Human Host: goat Form: lyophilized (IgG) Applications: Immunostaini ng of acetone-fixed frozen and formalin-fixed, paraffin-embedded tissue sections. HRP Conjugated 1 * 2 mL
Catalog # ICNA0855223
Supplier:  MP Biomedicals